Product Image
Thermo Scientific™
Data Outputs for Ultra-Low Temperature Freezers
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
195895RS-232 Output0/1 V
195896RS-232 Output0/5 V
195897RS-485 Output4-20mA
195898RS-485 Output0/1 V
195899RS-485 Output0/5 V
398213RS-232 Output, Factory Installed0/1 V
398214RS-485 Output, Factory Installed0/1 V
398215RS-485 Output, Factory Installed0/5 V
Catalog number 195895
Price (INR)
-
Type:
RS-232 Output
Voltage:
0/1 V
Integrate Thermo Scientific™ Data Outputs for Ultra-Low Temperature Freezers with a variety of ultra-low temperature freezers to increase functionality and compatibility.
RS-232:
  • RS-232 and 4-40mA analog outputs are standard on Thermo Scientific™ Forma™ 8600 Series freezers. These additional options are designed to meet specific needs.
  • RS-232 allows connection to a computer, recorder, or independent monitoring device RS-485 allows direct connection to the Model 1535 Monitor/Alarm System
Note:
RS-232 is not compatible with the Forma Model 1535 Monitor/Alarm System.
Specifications
For Use With (Equipment)Forma™ 8600, HERAfreeze™ HFC, Revco™ CxF, TSC Series
TypeRS-232 Output
Monitoring OptionsAlarm, Recorder
Voltage0/1 V
Unit SizeEach