Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals
Acros Organics

Zirconium standard solution, for AAS, 1mg/mL Zr in 5% HF, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
196471000
also known as 19647
100 mL
Catalog number 196471000
also known as 19647
HSN 28053000
Price (INR)
3,929.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
3,929.00
Each
Specifications
Chemical Name or MaterialZirconium standard solution
Name Note1 mg/ml Zr in 5% HF
PackagingPlastic bottle
CAS7440-67-7
Health Hazard 1GHS Signal Word: Danger
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for atomic adsorption spectroscopy
RUO – Research Use Only