Copper, 99%, powder, max. 106 μm (-140 mesh)
Copper, 99%, powder, max. 106 μm (-140 mesh)
Copper, 99%, powder, max. 106 μm (-140 mesh)
Acros Organics

Copper, 99%, powder, max. 106 μm (-140 mesh)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
25 g
500 g
Packaging:
Glass bottle
Catalog number 196570025
also known as 19657
HSN 74031900
Price (INR)
27,080.00
Each
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
27,080.00
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to red-brown
Appearance (Form)Powder
Assay>=98.5 % (Cu)
Average particle size10 to 28 µm
Particle size=<106 µm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only