Potassium iodate, 98%, extra pure
Potassium iodate, 98%, extra pure
Potassium iodate, 98%, extra pure
Acros Organics

Potassium iodate, 98%, extra pure

CAS: 7758-05-6 | IKO3 | 214 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 196741000
HSN 28299030
Price (INR)
7,220.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,220.00
Each
Chemical Identifiers
CAS7758-05-6
IUPAC Namepotassium iodate
Molecular FormulaIKO3
InChI KeyJLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES[K+].[O-][I](=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on drying=<0.5 % (105°C, 3 hrs)
Titration Iodometric>=97.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only