Tin(II) chloride dihydrate, 97%
Tin(II) chloride dihydrate, 97%
Tin(II) chloride dihydrate, 97%
Acros Organics

Tin(II) chloride dihydrate, 97%

CAS: 10025-69-1 | Cl2Sn·2H2O | 225.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
500 g
Catalog number 196800025
also known as 19680
HSN 28273910
Price (INR)
44,313.00
Each
Quantity:
2.5 kg
Request bulk or custom format
Price (INR)
44,313.00
Each
Chemical Identifiers
CAS10025-69-1
IUPAC Nameλ²-tin(2+) dihydrate dichloride
Molecular FormulaCl2H4O2Sn
InChI KeyFWPIDFUJEMBDLS-UHFFFAOYSA-L
SMILESO.O.[Cl-].[Cl-].[Sn++]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Appearance (Form)Crystalline powder or crystals and/or chunks
Titration Iodimetric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only