Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd
Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd
Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd
Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd
Acros Organics

Bis(triphenylphosphine)palladium(II) chloride, for analysis, 15% Pd

CAS: 13965-03-2 | C36H30Cl2P2Pd | 701.90 g/mol
Have Questions?
Catalog NumberQuantityPackaging
197320010
also known as 19732
1 gGlass bottle
Catalog number 197320010
also known as 19732
HSN 28439019
Price (INR)
12,830.00
Each
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,830.00
Each
Add to cart
Chemical Identifiers
CAS13965-03-2
IUPAC Namepalladium(2+) bis(triphenylphosphane) dichloride
Molecular FormulaC36H30Cl2P2Pd
InChI KeyYNHIGQDRGKUECZ-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Platinum (Pt)=<10 ppm
Copper (Cu)=<10 ppm
Gold (Au)=<10 ppm
Calcium (Ca)=<15 ppm
Appearance (Color)Yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only