2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%
Acros Organics

2,2-Dimethyl-1,3-dioxane-4,6-dione, 98%

CAS: 2033-24-1 | C6H8O4 | 144.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 198451000
HSN 29329900
Price (INR)
12,783.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,783.00
Each
Chemical Identifiers
CAS2033-24-1
IUPAC Name2,2-dimethyl-1,3-dioxane-4,6-dione
Molecular FormulaC6H8O4
InChI KeyGXHFUVWIGNLZSC-UHFFFAOYSA-N
SMILESCC1(C)OC(=O)CC(=O)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point92°C to 96°C
Titration with NaOH>=97.5 %
Appearance (Color)White to beige to light brown
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only