4-Phenoxybenzaldehyde, 98%
4-Phenoxybenzaldehyde, 98%
4-Phenoxybenzaldehyde, 98%
4-Phenoxybenzaldehyde, 98%
Acros Organics

4-Phenoxybenzaldehyde, 98%

CAS: 67-36-7 | C13H10O2 | 198.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 198930250
also known as 19893
HSN 29124999
Price (INR)
34,631.00
Each
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,631.00
Each
Add to cart
Chemical Identifiers
CAS67-36-7
IUPAC Name4-phenoxybenzaldehyde
Molecular FormulaC13H10O2
InChI KeyQWLHJVDRPZNVBS-UHFFFAOYSA-N
SMILESO=CC1=CC=C(OC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
GC>=97.5 %
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Refractive index1.6100 to 1.6120 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only