Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis)
Acros Organics

Yttrium(III) chloride hexahydrate, 99.9%, (trace metal basis)

CAS: 10025-94-2 | Cl36H2O | 303.35 g/mol
Have Questions?
Catalog NumberQuantityPackaging
19918050050 gGlass bottle
Catalog number 199180500
HSN 28469090
Price (INR)
17,470.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
17,470.00
Each
Chemical Identifiers
CAS10025-94-2
IUPAC Nameyttrium(3+) hexahydrate trichloride
Molecular FormulaCl3H12O6Y
InChI KeyIINACGXCEZNYTF-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Y+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystalline powder or crystals
Total trace metal impurities=<0.1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only