ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
Thermo Scientific™
ITEX Dynamic Headspace Tool for TriPlus™ RSH and TriPlus RSH SMART Autosamplers
Achieve highly sensitive determination of volatile organic compounds (VOCs) through simple, effective and automated dynamic headspace enrichment.
Have Questions?
Catalog NumberFor Use With (Equipment)
1R77010-1125TriPlus RSH and TriPlus RSH SMART Autosamplers
Catalog number 1R77010-1125
Price (INR)
-
For Use With (Equipment):
TriPlus RSH and TriPlus RSH SMART Autosamplers
Achieve ppt-level sensitivity for volatile organic compounds (VOCs) by integrating dynamic headspace enrichment into your automated sampling workflow for gas chromatography (GC). The ITEX-DHS tool enables a robust, easy-to-use, syringe-based headspace enrichment technique that offers high sorbent capacity and can be easily integrated into the Thermo Scientific™ TriPlus™ RSH and TriPlus RSH SMART autosamplers. Rapid and efficient enrichment followed by sample introduction are part of a fully automated workflow for extended unattended operations.
The ITEX-DHS technique uses a gas-tight syringe to collect the headspace and a microtrap inside the syringe needle to efficiently trap and concentrate VOCs through repeated strokes of the syringe. Scalable sensitivity can be achieved based on the number of strokes, making this a versatile approach for samples of both low and high concentrations.
For VOC analysis in many liquid and solid samples, ITEX can be a valuable alternative to classical enrichment techniques offering several benefits in terms of ease of use and robustness.
Easy, simple, robust and automated
  • Easy method set-up with few parameters to set
  • Simple hardware for an easy access to the trap
  • Excellent robustness for long-term operations
  • Closed system removing any risk of breakthrough
  • Independent trap desorption temperature and flow to maximize desorption process efficiency without impacting GC inlet settings
  • SMART gas-tight syringe to enable automatic usage traceability when the ITEX tool is used with the TriPlus RSH SMART autosampler.
  • Full control in the Thermo Scientific™ Chromeleon™ CDS to streamline the entire automated workflow
Versatile choices to meet your needs
  • ITEX Trap Carbosieve SIII 60/80 mesh
  • ITEX Trap Carboxen 1000 60/80 mesh
  • ITEX Trap Carbopack C 80/100 mesh
  • ITEX Trap Mix1 Tenax TA : Carboxen1000 (Bottom/Top = 1:1)
  • ITEX Trap Mix2 Carbopack C : Carbosieve SIII (Bottom/Top = 2:1)
  • ITEX Trap Mix3 Tenax GR : Carbosieve SIII (Bottom/Top = 1:1)
  • ITEX Trap Tenax GR 80/100 mesh
  • ITEX Trap Tenax TA 80/100 mesh
Specifications
DescriptionIncludes SMART gas-tight syringe, focusing trap, built-in trap heating and cooling fan and trap cleaning capability
For Use With (Application)Enrichment of volatile organic compounds (VOCs)
QuantityEach
For Use With (Equipment)TriPlus RSH and TriPlus RSH SMART Autosamplers
TypeAutosampler accessory
Unit SizeEach