Amberlyst™ 15, (wet) ion-exchange resin
Amberlyst™ 15, (wet) ion-exchange resin
Amberlyst™ 15, (wet) ion-exchange resin
Acros Organics

Amberlyst™ 15, (wet) ion-exchange resin

CAS: 9037-24-5 | MFCD00145841
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
Packaging:
Plastic bottle
Catalog number 202155000
HSN 98020000
Price (INR)
15,268.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
15,268.00
Each
Chemical Identifiers
CAS9037-24-5
Molecular FormulaMFCD00145841
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey-brown
Harmonic mean size0.600 to 0.850 mm
Particle size=<5.0 % (> 1.180 mm)
Total volume capacity>=1.70 eq/l (H)
Uniformity coefficient=<1.70
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only