Sodium iodide dihydrate, 99+%, for analysis
Sodium iodide dihydrate, 99+%, for analysis
Sodium iodide dihydrate, 99+%, for analysis
Sodium iodide dihydrate, 99+%, for analysis
Acros Organics

Sodium iodide dihydrate, 99+%, for analysis

CAS: 13517-06-1 | INa·2H2O | 185.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 202891000
also known as 20289
HSN 28276090
Price (INR)
8,190.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
8,190.00
Each
Add to cart
Chemical Identifiers
CAS13517-06-1
IUPAC Namesodium dihydrate iodide
Molecular FormulaH4INaO2
InChI KeyVEEIYHUOLCUPSA-UHFFFAOYSA-M
SMILESO.O.[Na+].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Magnesium (Mg)=<50 ppm
Lead (Pb)=<5 ppm
Appearance (Form)Adhering crystals
Nitrogen compounds (as N)=<0.002 %
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only