The Thermo Scientific SDS-Out SDS Precipitation Kit provides convenient removal of SDS from small-volume (0.1 to 10 mL) protein samples.FeaturesRead more
Have Questions?
Catalog Number
Quantity
20308
10 mL kit
Catalog number 20308
Price (INR)
29,903.00
Online offer
Ends: 31-Dec-2026
44,300.00
Save 14,397.00 (32%)
Each
Quantity:
10 mL kit
Request bulk or custom format
Price (INR)
29,903.00
Online offer
Ends: 31-Dec-2026
44,300.00
Save 14,397.00 (32%)
Each
The Thermo Scientific SDS-Out SDS Precipitation Kit provides convenient removal of SDS from small-volume (0.1 to 10 mL) protein samples.
Features of SDS-Out SDS Precipitation Reagent:
• Convenient removal of SDS from microliter-volume samples • Use for samples containing 0.1 to 1% SDS • Minimal dilution of small-volume samples • Protein recovery is 90 to 100% (based on tests with BSA, cytochrome C, soybean trypsin inhibitor, ovalbumin, ribonuclease A, myoglobin and human IgG)
The simple reagent and kit effectively remove excess SDS from small protein samples. Sodium dodecyl sulfate (SDS) is a detergent typically used in cell lysis reagents and other protein solubilization buffers. Traditional methods for SDS removal include prolonged dialysis, anion exchange chromatography, affinity columns or and acetone precipitation. By comparison, the SDS-Out Precipitation Reagent is relatively easy to use and provides excellent protein recovery.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypePrecipitation, Filtration, n/a
FormatKit, Microcentrifuge, Spin Column
Purification TargetDetergent Removal
Quantity10 mL kit
Sufficient For200 mL Protein Solution
Product LineSDS-Out
Product TypeSDS Precipitation Kit
Unit SizeEach
Contents & Storage
Sufficient For: Precipitation of SDS from 200 mL of protein solution • SDS Precipitation Reagent, 10 mL • Spin Cup Columns, 12 columns • Collection Tubes, 12 tubes