(1S)-(-)-Verbenone, 94%
(1S)-(-)-Verbenone, 94%
(1S)-(-)-Verbenone, 94%
Acros Organics

(1S)-(-)-Verbenone, 94%

CAS: 1196-01-6 | C10H14O | 150.22 g/mol
Have Questions?
Catalog NumberQuantityPackaging
203630500
also known as 20363
50 gGlass bottle
Catalog number 203630500
also known as 20363
HSN 29142990
Price (INR)
19,624.00
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
19,624.00
Each
Chemical Identifiers
CAS1196-01-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.4950 to 1.4970 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=92 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only