Diethylstilbestrol, 99%
Diethylstilbestrol, 99%
Diethylstilbestrol, 99%
Acros Organics

Diethylstilbestrol, 99%

CAS: 56-53-1 | C18H20O2 | 268.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 204130010
also known as 20413
HSN 98020000
Price (INR)
6,912.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
6,912.00
Each
Chemical Identifiers
CAS56-53-1
IUPAC Name4-[(3E)-4-(4-hydroxyphenyl)hex-3-en-3-yl]phenol
Molecular FormulaC18H20O2
InChI KeyRGLYKWWBQGJZGM-ISLYRVAYSA-N
SMILESCCC(=C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Melting point169°C to 175°C
HPLC>=98.5 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only