Ammonium oxalate monohydrate, 99.5%, for analysis
Ammonium oxalate monohydrate, 99.5%, for analysis
Ammonium oxalate monohydrate, 99.5%, for analysis
Acros Organics

Ammonium oxalate monohydrate, 99.5%, for analysis

CAS: 6009-70-7 | C2H8N2O4·H2O | 142.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
500 g
Packaging:
Plastic bottle
Catalog number 206270010
HSN 29171190
Price (INR)
18,329.00
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
18,329.00
Each
Chemical Identifiers
CAS6009-70-7
IUPAC Nameoxalic acid diamine hydrate
Molecular FormulaC2H10N2O5
InChI KeyMSMNVXKYCPHLLN-UHFFFAOYSA-N
SMILESN.N.O.OC(=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Sulfate (SO4)=<50 ppm
Titration with KMnO499.4 to 101.0 %
Chloride (Cl)=<20 ppm
Iron (Fe)=<3 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only