Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade
Acros Organics

Cesium chloride, 99.99%, (trace metal basis), for spectroscopy, optical grade

CAS: 7647-17-8 | ClCs | 168.36 g/mol
Have Questions?
Catalog NumberQuantityPackaging
206320050
also known as 20632
5 gGlass bottle
Catalog number 206320050
also known as 20632
HSN 28273990
Price (INR)
4,071.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,071.00
Each
Chemical Identifiers
CAS7647-17-8
IUPAC Namecaesium(1+) chloride
Molecular FormulaClCs
InChI KeyAIYUHDOJVYHVIT-UHFFFAOYSA-M
SMILES[Cl-].[Cs+]
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble matter=<25 ppm (in water)
UVat 280 nm A: =<0.02
Appearance (Form)Crystalline powder or crystals
Purity>=99.94 % (metals basis)
UV(50 % in water) 1 cm cell
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis, For UV/VIS spectroscopy
RUO – Research Use Only