Murexide, ACS reagent
Murexide, ACS reagent
Murexide, ACS reagent
Acros Organics

Murexide, ACS reagent

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass Bottle
Catalog number 206890250
HSN 29335990
Price (INR)
19,363.00
Each
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
19,363.00
Each
Chemical Identifiers
CAS3051-09-0
IUPAC Nameammonium 2,4,6-trioxo-5-[(2,4,6-trioxo-1,3-diazinan-5-ylidene)amino]-1,3-diazinan-5-ide
Molecular FormulaC8H8N6O6
InChI KeyPCCZWUCAYILEMX-UHFFFAOYSA-O
SMILES[NH4+].O=C1NC(=O)[C-](N=C2C(=O)NC(=O)NC2=O)C(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Bordeaux to red-purple
Infrared spectrumConforms
Appearance (Form)Powder
Suitability(as complexom. indicator) Passes test
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only