Procaine hydrochloride, 99%
Procaine hydrochloride, 99%
Procaine hydrochloride, 99%
Acros Organics

Procaine hydrochloride, 99%

CAS: 51-05-8 | C13H21ClN2O2 | 272.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number 207315000
also known as 20731
HSN 29420090
Price (INR)
24,249.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
24,249.00
Each
Chemical Identifiers
CAS51-05-8
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<0.5 % (1 g, 105°C)
Appearance (Form)Crystalline powder or crystals
Titration with NaOH98.5 to 101.5 %
pH5.0 to 6.5 (2 % w/v in water)
Appearance (Color)Colorless to white to almost white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only