Sodium chloride, 99.5%, for analysis
Sodium chloride, 99.5%, for analysis
Sodium chloride, 99.5%, for analysis
Acros Organics

Sodium chloride, 99.5%, for analysis

Sodium chloride, 99.5%, ClNa, CAS Number-7647-14-5, sodium chloride nacl, salt, common salt, rock salt, halite, purex, table salt, sodium chloride, saline, dendritis, 1kg, 99.4% min. (Argentometry), 10ppm max., 1461.0 deg.C, 50ppm max., 20ppm max., CHEBI:26710, White, 231-598-3, 13,280, 58.44 | CAS: 7647-14-5 | ClNa | 58.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 kg
1 kg
5 kg
Packaging:
View all
Plastic Drum
Catalog number 207790250
also known as 20779
HSN 25010090
Price (INR)
45,757.00
Each
Quantity:
25 kg
Packaging:
Plastic Drum
Request bulk or custom format
Chemical Identifiers
CAS7647-14-5
IUPAC Namesodium chloride
Molecular FormulaClNa
InChI KeyFAPWRFPIFSIZLT-UHFFFAOYSA-M
SMILES[Na+].[Cl-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Heavy metals (as Pb)=<5 ppm
Total nitrogen (as N)=<0.001 %
Barium (Ba)=<10 ppm
Potassium (K)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer
RUO – Research Use Only