Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Sodium carbonate, 99.8%, for analysis, anhydrous, powder
Acros Organics

Sodium carbonate, 99.8%, for analysis, anhydrous, powder

CAS: 497-19-8 | CNa2O3 | 105.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
5 kg
Packaging:
View all
Plastic Bottle
Catalog number 207810010
also known as 20781
HSN 28362090
Price (INR)
8,637.00
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS497-19-8
IUPAC Namedisodium carbonate
Molecular FormulaCNa2O3
InChI KeyCDBYLPFSWZWCQE-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HCl>=99.8 % (On dried substance)
Total sulfur (as SO4)=<0.003 %
Chloride (Cl)=<10 ppm
Aluminium (Al)=<10 ppm
Potassium (K)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Biological buffer
RUO – Research Use Only