Bis(cyclopentadienyl)zirconium chloride hydride, 95%
Bis(cyclopentadienyl)zirconium chloride hydride, 95%
Bis(cyclopentadienyl)zirconium chloride hydride, 95%
Acros Organics

Bis(cyclopentadienyl)zirconium chloride hydride, 95%

CAS: 37342-97-5 | C10H11ClZr | 257.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
25 g
Packaging:
Glass bottle
Catalog number 207890050
also known as 20789
HSN 29319090
Price (INR)
17,351.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
17,351.00
Each
Chemical Identifiers
CAS37342-97-5
IUPAC Nameλ²-zirconium(2+) bis(cyclopenta-2,4-dien-1-ide) chloride hydride
Molecular FormulaC10H11ClZr
InChI KeyVPQMECJLNPUNGN-UHFFFAOYSA-M
SMILES[H-].[Cl-].[Zr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric>=94 %
Appearance (Color)White to beige
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only