Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™
Acros Organics

Cyclopentylmagnesium chloride, 2M solution in diethyl ether, AcroSeal™

CAS: 32916-51-1 | C5H9ClMg | 128.88 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 208468000
also known as 20846
HSN 29319090
Price (INR)
67,251.00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (INR)
67,251.00
Each
Chemical Identifiers
CAS32916-51-1
IUPAC Namechloro(cyclopentyl)magnesium
Molecular FormulaC5H9ClMg
InChI KeyGROGEABZCKQTJW-UHFFFAOYSA-M
SMILESCl[Mg]C1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Grignard1.9 to 2.2 M
Appearance (Color)Clear to hazy yellow to grey to brown
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only