2-(Methylthio)ethanol, 99%
2-(Methylthio)ethanol, 99%
2-(Methylthio)ethanol, 99%
Acros Organics

2-(Methylthio)ethanol, 99%

CAS: 5271-38-5 | C3H8OS | 92.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 210101000
also known as 21010
HSN 29309099
Price (INR)
34,986.00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
34,986.00
Each
Chemical Identifiers
CAS5271-38-5
IUPAC Name2-(methylsulfanyl)ethan-1-ol
Molecular FormulaC3H8OS
InChI KeyWBBPRCNXBQTYLF-UHFFFAOYSA-N
SMILESCSCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4934 to 1.4954 (20°C, 589 nm)
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only