Try new Gibco Insulin-Transferrin-Selenium Select supplements, an animal-origin free formulation. Available in ITS-G & ITS-X. Learn more >
DMEM/F-12, no phenol red
DMEM/F-12, no phenol red
DMEM/F-12, no phenol red
Gibco™

DMEM/F-12, no phenol red

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many differentRead more
Have Questions?
Catalog NumberQuantity
21041025500 mL
Catalog number 21041025
Price (INR)
3,910.00
Online offer
Ends: 31-Dec-2026
5,585.00
Save 1,675.00 (30%)
Each
Quantity:
500 mL
Customize this product
Price (INR)
3,910.00
Online offer
Ends: 31-Dec-2026
5,585.00
Save 1,675.00 (30%)
Each

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.


This DMEM/F-12 is modified as follows:
WithWithout
• L-glutamine• HEPES
 • Phenol Red


The complete formulation is available.

Using DMEM/F-12
DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell LineMDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeDMEM (Dulbecco's Modified Eagle Medium)/F-12
Quantity500 mL
Shelf Life12 Months From Date of Manufacture
Shipping ConditionRoom Temperature
ClassificationAnimal Origin-free
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Sodium Pyruvate
Without AdditivesNo Phenol Red, No HEPES
Unit SizeEach
Contents & Storage
Storage conditions: 2-8° C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Do you offer DMEM/F12 medium without folic acid?

We do not offer DMEM/F12 medium without folic acid as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I understand that some media are worse than others for fluorescence imaging. How do I choose?

Most media contain phenol red, which can quench fluorescent dyes in the visible wavelengths. Most media also contain autofluorescent components, such as riboflavin, which can reduce signal-to-background. We offer FluoroBrite DMEM and HEPES-based Live Cell Imaging Solution, which have been optimized for fluorescent imaging. We also offer a number of media without phenol red. But if none of these are reasonable options for your experiment, then we also offer BackDrop Background Suppressor ReadyProbes Reagent, which can be added to quench media autofluorescence.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Should I be concerned about phenol red in my media when labeling my live cells with fluorescent dyes?

Some cell types accumulate phenol red, and this can pose a problem in the use of many fluorescent probes. Phenol red can quench visible-wavelength dyes and, although phenol red is non-fluorescent, various impurities may be fluorescent. We have many phenol red-free media to choose from. Our Live Cell Imaging Solution (HEPES-based) and our FluoroBrite DMEM have been optimized to be phenol red-free as well as to be non-autofluorescent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

Citations & References (1)

Citations & References
Abstract
Study of neurotoxic intracellular calcium signalling triggered by amyloids.
Authors:Villalobos C, Caballero E, Sanz-Blasco S, Núñez L
Journal:Methods Mol Biol
PubMed ID:22528098
Neurotoxicity in Alzheimer's disease (AD) is associated to dishomeostasis of intracellular Ca(2+) induced by amyloid ß peptide (Aß) species. Understanding of the effects of Aß on intracellular Ca(2+) homeostasis requires preparation of the different Aß assemblies including oligomers and fibrils and the testing of their effects on cytosolic and mitochondrial ... More