Product Image
Gibco™

DMEM/F-12, no phenol red

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many differentRead more
Have Questions?
Catalog NumberQuantity
21041033
also known as 21041-033
10 x 500 mL
Catalog number 21041033
also known as 21041-033
Price (INR)
-
Quantity:
10 x 500 mL
Customize this product

DMEM/F-12 (Dulbecco's Modified Eagle Medium/Nutrient Mixture F-12) is a widely used basal medium for supporting the growth of many different mammalian cells. Cells successfully cultured in DMEM/F-12 include MDCK, glial cells, fibroblasts, human endothelial cells, and rat fibroblasts. We offer a variety of DMEM/F-12 modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This DMEM/F-12 is modified as follows:

The complete formulation is available.

Using DMEM/F-12

DMEM/F-12 is a 1:1 mixture of DMEM and Ham's F-12. This formulation combines DMEM's high concentrations of glucose, amino acids and vitamins with F-12's wide variety of components. DMEM/F-12 contains no proteins, lipids, or growth factors. Therefore, DMEM/F-12 may require supplementation, commonly with 10% Fetal Bovine Serum (FBS). DMEM/F-12 uses a sodium bicarbonate buffer system and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeDMEM/F-12
Quantity10 x 500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
Serum LevelStandard Serum Supplementation
SterilitySterile-filtered
With AdditivesHigh Glucose, Glutamine, Sodium Pyruvate
Without AdditivesNo HEPES, No Phenol Red
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Frequently asked questions (FAQs)

Do you offer DMEM/F12 medium without folic acid?

We do not offer DMEM/F12 medium without folic acid as a standard catalog product. However, our Customs group can make it for you as a custom media. If you are interested, please complete this Custom order inquiry form PDF (https://www.thermofisher.com/content/dam/LifeTech/global/applied-sciences/pdfs/Bioproduction/Gibco-Custom-Media-Buffers-Request-Form.pdf) and email it to custommedia@thermofisher.com. Our Customs group will review your requirements and get back to you within a couple of days with pricing, ordering information, and lead time. After that, you have the choice of proceeding with an “official” order or declining.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Do you offer media without phenol red (phenol red-free media)?

Yes, please visit this page for a list of our phenol red-free media (http://tools.thermofisher.com/content/sfs/brochures/Phenol_RedFree_Media_Glance.pdf).

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.