Orion™ 2100 Series pH ORP and Conductivity Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2100 Series pH ORP and Conductivity Analyzer

Measure pH, ORP and conductivity accurately and reliably in the harshest environments with our pH ORP and conductivity analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeChannels
2104CD2SSAnalyzer, UPW Conductivity Kit2
2100CD2Conductivity Module
2100PH2pH/ORP Module
2100PMKPipe Mounting Kit
2100SMKSample Panel Mounting Kit
2102PH1SCAnalyzer with UPW pH Kit1
2102PH2pH/ORP Analyzer2
2102PHXAnalyzer with ORP Kit1
2104CDConductivity Analyzer2
Catalog number 2104CD2SS
Price (INR)
-
Type:
Analyzer, UPW Conductivity Kit
Channels:
2
Measure pH/ORP and conductivity accurately and reliably in the harshest environments with the Thermo Scientific™ Orion™ 2100 Series pH ORP and Conductivity Analyzer. Designed for the highest accuracy and performance, this analyzer has flexible measurement parameters for single, dual or combined pH ORP. Conductivity inputs are offered with optional digital communications.

Features:

  • Single channel or dual channel configurations
  • Optional digital communication boards
  • Provides cation and ammonia/ETA compensation for customizing to your plant's requirement

2100 Series pH Analyzer Consumables Ordering Information:

  • pH Consumables Kit — pH 4.01, 7.00 and 10.01 Buffers and Process Thermo Scientific™ Ross™ Electrode Filling Solution, Cat. No. 2001CP
  • pH 4.01 Buffer, 475mL, Cat. No. 910104
  • pH 7.00 Buffer, 475mL, Cat. No. 910107
  • pH 10.01 Buffer, 475mL, Cat. No. 910110
  • Process Ross Electrode Filling Solution, 5 x 60mL, Cat. No. 2001FS
  • Process Ross Electrode Filling Solution, 4 x 120mL, Cat. No. 2001F4
  • pH Electrode Cleaning Solution A for Protein Removal, 4 x 30mL, Cat. No. 900021
  • pH Electrode Cleaning Solution B for Bacteria Removal, 4 x 60mL, Cat. No. 900022
  • pH Electrode Cleaning Solution C for General Cleaning, 4 x 30mL, Cat. No. 900023
  • pH Electrode Cleaning Solution D for Oil and Grease Removal, 4 x 60mL, Cat. No. 900024

2100 Series Conductivity Analyzer Consumables Ordering Information:

  • 111.9mS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011005
  • 12.9mS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011006
  • 1413μS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011007
  • 100μS/cm Conductivity Standard, 5 x 60mL, Cat. No. 011008
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy±0.01
Accuracy (Resistivity)0.5% ± 1 digit
Accuracy (Temperature)±0.5°C
Alarm Output3 ea.
Amperage5A
Analog Outputs2 ea.
Case MaterialValox 364
Certifications/ComplianceCE,CSA, FCC Class A limits
Channel 1Conductivity
Channel 2Conductivity
Channels2
Communication DetailsModBus with additional Module
Current Loops4(0) to 20 mA
Description2104CD Dual Channel Analyzer and Ultra Pure Water (UPW) Conductivity Kit
Display TypeCustom LCD, backlit with scrolling text
Enclosure RatingNEMA 4X IP 65
Frequency0 to 60 Hz
Height (Metric)144 mm
Includes2 x 2002SS conductivity cells, 2 flow cells (2002FC) and 100 uS/cm conductivity standard (011008)
Keypad TypeMembrane
Length (Metric)186 mm
Max. Relative Humidity5 to 95%, non-condensing
Mounting OptionsPipe, Panel or Wall Mount
Range (Resistivity)2ohm to 100Mohm
Range (mV)±1999mV
Resolution0.1; 0.01
Resolution (Resistivity)2ohms/cm
Resolution (Temperature)0.1°C
Resolution (mV)1mV
Temperature (English) Operating41°F to 113°F
USB InterfaceNo
Voltage85/132 VAC, 170/264 VAC
Width (Metric)144 mm
Temperature (Metric) Operating5°C to 45°C
TypeAnalyzer, UPW Conductivity Kit
Unit SizeEach