Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Actual product may vary
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure
Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Nalgene™ Square Wide-Mouth PPCO Bottles with Closure

Simplify cleaning, sample filling and retrieval with these space-efficient, wide-mouth PPCO bottles, which are contact-clear, autoclavable and resistant to most acids, alcohols and bases.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2110-0006175 mL38 mm
2110-000260 mL28 mm
2110-0008250 mL43 mm
2110-0016500 mL53 mm
2110-00321 L63 mm
Catalog number 2110-0006
HSN 39233090
Price (INR)
20,381.00
Case
Capacity (Metric):
175 mL
Closure Size:
38 mm
Price (INR)
20,381.00
Case

Save valuable lab space with the square design of Thermo Scientific™ Nalgene™ Wide-Mouth Square Polypropylene Copolymer Bottles. Wide-mouth design allows easy cleaning, sample filling and retrieval. Made from only high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables.

Features:
  • Resistant to acids, alcohols and bases
  • Contact-clear bottle provides visible liquid level
  • Square design provides a large area for labeling
  • Linerless polypropylene closures provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Autoclavable
  • Leakproof† when used with Nalgene closures
  • Manufactured from non-toxic materials

Note:

Completely disengage threads or remove cap before autoclaving.

.

Recycling Information:
Nalgene reusable polypropylene(PP) and polypropylene copolymer (PPCO) bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  These bottles and their packaging are recyclable in many communities. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene PP and PPCO bottles if services are available in your community.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity6 oz., 175 mL
Capacity (Metric)175 mL
Capacity (English)6 oz.
ColorTranslucent
DescriptionWide-Mouth Square Bottle
Diameter (English)2 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
MaterialPPCO
No. per Case72
No. per Pack12
SterilityNon-sterile
Closure ColorNatural, translucent
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
GraduatedNo
MouthWide
ShapeSquare
Height (Metric)102 mm
Product TypeSquare Wide Mouth Bottle
Width (English)2.05 in.
Height (English)4 in.
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size38 mm
Diameter (Metric)51 mm
Closure TypeScrew Cap
Through the Closure DispensingNo
AutoclavableAutoclavable
Unit SizeCase

Frequently asked questions (FAQs)

Are the Nalgene PPCO media bottles/tubes able to be irradiated?

PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.

Why are your Nalgene containers and caps always different materials?

In order to function as a leakproof system without a liner (one of the key advantages of our laboratory and packaging products), the surfaces of the container and closure that meet and push together when tightened must be dissimilar materials with different hardnesses. Even our polypropylene copolymer (PPCO) bottles with polypropylene homopolymer (PP) bottles are different enough to function properly. On some larger carboy products, this difference cannot be sufficiently maintained so a pliable thermoplastic elastomer (TPE) gasket is added for proper leakproof seal function.