Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Actual product may vary
Thermo Scientific™
Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for Orion™ 2110XP Ammonia Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2110XPSKService Kit
210056Reference Electrode Half-Cell
211050XPAmmonia Consumables Kit
211050XPFReagentless Consumables Kit
2110XPCALAmmonia Calibration Kit
2110XPEKAmmonia Electrode Kit
2110XPEPElectronics Faceplate
Catalog number 2110XPSK
Price (INR)
-
Type:
Service Kit
Keep needed supplies and service items in stock to avoid lost productivity with Thermo Scientific™ Consumables and Services Kits for the Thermo Scientific™ Orion™ 2110XP Ammonia Analyzer.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Accuracy (mV)±(0.5 mV + 0.1%)
Description2110XP Service Kit
Includeselectronics faceplate (2110XPEP), air pump assembly (2100PA), regulator assembly (2100RG), 2 electrode cables with gland assemblies for sensing and reference electrodes (2100EC), power supply assembly (2100PS) and fuse kit assembly (2100FK115 and 2100FK230)
Response TimeInitial response within 15 seconds, reach 90% of reading within 1 minute of injecting a standard solution
Temperature (Metric) Operating5°C to 45°C (Sample)
TypeService Kit
Unit SizeEach