Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Actual product may vary
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers
Thermo Scientific™

Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzers

Adapt your sodium analyzer to specific needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for Orion™ 2111XP Sodium Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
211150XPFReagentless Cation/High Acid Kit
210045Sodium ISE
211150XPCation/High Acid Consumables
211160XPAmmonia Consumables Kit
211160XPFReagentless Ammonia Kit
211192XPDIPA Consumables Kit
211192XPFReagentless DIPA Consumables Kit
2111XAAmmonia Application Package
2111XCCation/High Acid Package
2111XDDIPA Application Package
2111XPCALSodium Calibration Kit
2111XPEKElectrode Kit
2111XPEPElectronics Faceplate
Catalog number 211150XPF
Price (INR)
-
Type:
Reagentless Cation/High Acid Kit
Adapt your sodium analyzer to your specific analytical needs with Thermo Scientific™ Application Packages, Consumables and Replacement Parts for the Thermo Scientific™ Orion™ 2111XP Sodium Analyzer.
Warranty: Out of box
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Case MaterialValox 264
Certifications/Compliance211150XPF
DescriptionReagent-less Cation/High Acid Application Consumables Kit
Detection Limits0.1 ppb
Display Type211150XPF
Flow Rate40 mL/min.
For Use With (Equipment)Orion™ 2111XP Sodium Analyzer
Includes12 x 2L empty bottles for ammonia reagent, diffusion tubing (181160), sodium calibration standards kit (181140), sodium electrode (210045), reference electrode (210056), 5 x 2 oz. reference filling solution (181073), O-ring kit, check valve, box of 0.1mL
Inlet Pressure8 to 100 psig
OutputsAnalog: 3; Alarm: 3; Relay: 3
Relay DetailsMax Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes
Sample Inlet1/4 in. NPTF tube fitting
Sample Temperature5 to 45 °C
Voltage85/132 VAC, 170/264 VAC
Temperature (Metric) Operating5°C to 45°C (Sample)
TypeReagentless Cation/High Acid Kit
Unit SizeEach