Orion™ 2111LL Low Level Sodium Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2111LL Low Level Sodium Analyzer

Protect your turbine against destructive sodium levels, including trace amounts, with the reliable and easy to use Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberIncludes
2111LLGGrab Sampler Package
2111LLInstrument Only Package
2111LLENProtective Enclosure Package
2111LLENGGrab Sampler/Enclosure Package
Catalog number 2111LLG
Price (INR)
-
Includes:
Grab Sampler Package
Protect your turbine against destructive sodium levels, including trace amounts, with the reliable and easy to use Thermo Scientific™ Orion™ 2111LL Low Level Sodium Analyzer. Available in several configurations including the analyzer instrument only, or with a protective enclosure.

Features:

  • Resistant to drift with Orion™ Ross™ UltraTM electrode technology
  • Reduces hazardous waste
  • Easy to read backlit display for measurement visibility from a distance
  • No moving parts with the pum-less passive diffusion reagent system
  • Rapid response to changes and ultra low level detection with proprietary flow cell

Ordering information:

  • All low level sodium monitors come with step-by-step instructions for quick installation and start-up.
  • Available in several configurations including the analyzer instrument only or with a protective enclosure.
Specifications
Accuracy±5% or 0.01 ppb, whichever is greater
Accuracy (Temperature)±0.5°C
Accuracy (mV)± (0.5 mV + 0.1%)
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channel 1Sodium
Channel 2Optional ph/ORP or Conductivity
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2111LL Low Level Sodium Analyzer and Grab Sampler
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40mL/min.
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
IncludesGrab Sampler Package
Inlet Pressure8 to 100psig
Keypad TypeMembrane
Length (English)11 in.
Length (Metric)27 cm
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Range0.001ppb to 10ppm
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Resolution1; 2; 3; or 4 significant digits
Response Time95% within 2 minutes of injecting a standard solution
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Sample Temperature5 to 45°C
Voltage85/132 VAC, 170/264 VAC
Weight (English)50 lb.
Weight (Metric)22.7 g
Width (English)17 in.
Width (Metric)45 cm
Temperature (Metric) Operating5°C to 35°C
TypeSodium Analyzer
Unit SizeEach