Streptavidin Protein, HRP
Thermo Scientific™
Streptavidin Protein, HRP
Thermo Scientific™ Pierce™ Streptavidin, Horseradish Peroxidase Conjugate includes streptavidin in a purified form conjugated to peroxidase for substrate-based detection.Streptavidin, HorseradishRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
211275 mg
211261 mg
211242 mg
Catalog number 21127
Price (INR)
58,393.00
Online offer
Ends: 31-Dec-2026
97,321.00
Save 38,928.00 (40%)
Each
Quantity:
5 mg
Request bulk or custom format
Price (INR)
58,393.00
Online offer
Ends: 31-Dec-2026
97,321.00
Save 38,928.00 (40%)
Each
Thermo Scientific™ Pierce™ Streptavidin, Horseradish Peroxidase Conjugate includes streptavidin in a purified form conjugated to peroxidase for substrate-based detection.
Streptavidin, Horseradish Peroxidase Conjugate enables detection of biotinylated antibodies and other probes in a variety of standard assay methods, including western blotting, ELISA, immunohistochemistry, and fluorescence imaging. Streptavidin, Horseradish Peroxidase conjugate is supplied as lyophilized powder in phosphate-based buffer for immediate reconstitution with water.
Streptavidin is a biotin-binding protein originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Pierce streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Recommended for use in ELISA (0.1 μg/mL), immunohistochemistry (1-10 μg/mL), and western blot (2 μg/mL) applications.
Related products
  • Pierce™ Streptavidin (Cat. No. 21122)
  • Pierce™ Streptavidin, Alkaline Phosphatase Conjugated (Cat. No. 21324)
  • Pierce™ Streptavidin, Maleimide-Activated (Cat. No. 21102)
  • Pierce™ Streptavidin, Hydrazide-Activated (Cat. No. 21120)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Product TypeStreptavidin Conjugate
Quantity5 mg
Shipping ConditionAmbient Temperature
Target MoleculeBiotin
ConjugateHRP
FormSolid
Product LinePierce
Unit SizeEach
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.