Bacto™ Tryptose
Gibco™

Bacto™ Tryptose

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21170910 kg
211713500 g
Catalog number 211709
Price (INR)
-
Quantity:
10 kg

Bacto Tryptose is a mixed enzymatic hydrolysate with distinctive nutritional properties. The digestive process of tryptose results in assorted peptides of higher molecular weight suitable for long-chain amino acid requirements.

Features:

  • Reported as beneficial for cell culture applications
  • Animal origin (AO) nutritional supplement

Applications of Bacto Tryptose include:

  • Tryptose is very useful for cultivation of streptococci, pneumococci, meningococci, and other fastidious organisms.
  • Tryptose Peptone is often used as a biomass enhancer for recombinant Escherichia coli production.
Specifications
For Use With (Application)Cell Culture
Product LineBacto
Quantity10 kg
SourceAnimal Origin
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.