Orion™ 2117LL Low-Level Chloride Analyzer
Actual product may vary
Orion™ 2117LL Low-Level Chloride Analyzer
Orion™ 2117LL Low-Level Chloride Analyzer
Thermo Scientific™
Orion™ 2117LL Low-Level Chloride Analyzer
Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
2117LL230Chloride Analyzer230 V
2117LLChloride Analyzer110 V
2117LLENChloride Analyzer, Enclosure110 V
2117LLEN230Chloride Analyzer, Enclosure230 V
Catalog number 2117LL230
Price (INR)
-
Type:
Chloride Analyzer
Voltage:
230 V
Get continuous online monitoring and control of steam cycle and boiler chemistry with the Thermo Scientific™ Orion™ 2117LL Low-Level Chloride Analyzer, for use with power generation applications requiring chloride measurement at trace levels.
Without accurate and real-time monitoring of chloride, the risk of corrosion fatigue, stress corrosion cracking and pitting can create significant costs in maintenance and downtime. Early detection of chloride ingress from feedwater helps your plant maintain the optimal balance of water purification with ease and peace of mind.
Features:
  • Detects chloride in real-time, providing accurate and reliable results
  • Provides high performance results with confidence
  • Includes unique electrode technology for stability and a virtually maintenance-free fluidic design
Benefits:
  • Calibration record is maintained for up to 60 days between reagent changes.
  • System is one of the only real-time monitors on the market capable of reading down to 5ppb.
  • Long-life, solid-state sensor technology offers stability and high performance.
  • No moving parts means simple operation and minimal maintenance.
  • Measurements are repeatable and verifiable.
Ordering information:
The chloride analyzer is available in several configurations including the analyzer instrument only or with a protective enclosure.
Specifications
DescriptionLow Level Chloride Analyzer, 230V
Voltage230 V
TypeChloride Analyzer
Unit SizeEach