Kits and Accessories for Orion™ 2117XP Chloride Analyzers
Actual product may vary
Thermo Scientific™
Kits and Accessories for Orion™ 2117XP Chloride Analyzers
Keep chloride analyzers well-supplied and ready for use with the Thermo Scientific™ Kits and Accessories for Orion™ 2117XP Chloride Analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2117XPEKChloride Electrode Kit
211750XPChloride Consumables Kit w/ Reagents
211750XPFChloride Consumables Kit w/o Reagents
2117RBAReagent Bottle Adapter
2117XPCALCalibration Kit
2117XPEPElectronics Faceplate
2117XPSKService Kit
Catalog number 2117XPEK
Price (INR)
-
Type:
Chloride Electrode Kit
Keep chloride analyzers well-supplied and ready for use with the Thermo Scientific™ Kits and Accessories for Orion™ 2117XP Chloride Analyzers. Choose from the reagent-less consumables kit, reagent bottle adapter, chloride calibration kit or electrode kit.
Warranty: Out of box
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
For Use With (Equipment)Orion™ 2117XP Chloride Analyzers
TypeChloride Electrode Kit
Unit SizeEach