Bacto™ Peptone
Gibco™

Bacto™ Peptone

Bacto Peptone is an animal-origin (AO) enzymatic digest of bovine and porcine animal proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
21183010 kgPail
211677500 gBottle
211677-44 x 500 gBottle(s)
2118202 kgPail
Catalog number 211830
Price (INR)
274,241.00
Each
Quantity:
10 kg
Format:
Pail
Price (INR)
274,241.00
Each
Bacto Peptone is an animal-origin enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents, and a unique amino acid content that makes this peptone a strong nitrogen source. Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisiae, Escherichia coli, Pasteurella multocida, Vibrio cholerae, and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.

Features:

  • A strong nitrogen source
  • Unique amino acid content
  • High concentration of complex constituents

Applications of Bacto Peptone include:

  • Vaccines for animal health applications
  • Production of biotherapeutics
  • Mammalian cell culture
  • Microbial fermentation
Specifications
Cell TypeBacteria
Certifications/ComplianceLUO, MUO
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
FormatPail
Product LineBacto
Quantity10 kg
Shelf Life1825 Days
SourceAnimal origin
FormPowder
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.