Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers
Actual product may vary
Thermo Scientific™
Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers
Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2118XPEKElectrode Kit
211850XPO2 Scavenger Consumables Kit
211850XPFReagentless O2 Scavenger Kit
2118CBBlock Assembly
2118FCFlow-cell Assembly
2118MCSpool Assembly
2118MVMicro Valve Assembly
2118RBAAdapter Assembly
2118XPEPElectronics Faceplate
2118XPSKService Kit
21180KO-ring Kit
2118FPPanel Assembly
2118PNNeedle Assembly
2118CVCalibration Valve Assembly
Catalog number 2118XPEK
Price (INR)
-
Type:
Electrode Kit
Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers. Choose from options including: oxygen scavenger consumables kit; reagent-less oxygen scavenger consumables kit; fluid connector block assembly with thumb screw and injection port; flow cell assembly with electrode holder, mounting bracket and O-rings; and mixing coil-tubing spool assembly.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description2118XP Electrode Kit - includes iodide electrode, reference electrode and 2 oz. reference electrode filling solution
For Use With (Equipment)Orion™ 2118XP Oxygen Scavenger Analyzers
IncludesIodide electrode, reference electrode and 2 oz. reference electrode filling solution
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeElectrode Kit
Unit SizeEach