Bacto Proteose Peptones are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parametersRead more
Have Questions?
Change view
Catalog Number
Quantity
212010
10 kg
211684
500 g
2 Options
Catalog number 212010
Price (INR)
595,879.00
Each
Quantity:
10 kg
Price (INR)
595,879.00
Each
Bacto Proteose Peptones are enzymatic digests of protein. Studies of peptic digests of animal tissue prepared under varying digestion parameters demonstrated that no single peptone is the most suitable nitrogen source for every microbiological application. These findings led to the development of Bacto Proteose Peptone, Bacto Proteose Peptone No. 2, and Bacto Proteose Peptone No. 3. Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone.
• Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins • Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media • Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media • Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone. It offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
We also offer alternatives to Bacto Proteose Peptones for scale-up to production applications, including BiTek Proteose Peptone.
Specifications
Certifications/ComplianceLUO, MUO
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
FormatPail
Product LineBacto
Quantity10 kg
Shelf Life1825 Days
SourceAnimal Origin
FormDehydrated, free-flowing granules
Product TypeBacterial Media
pH6.6–7.6 (1.0% solution at 25°C)
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.