Product Image
Thermo Scientific™
Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits
Accessories and kits for the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer include a consumables kit, calibration kit, carrying case, electrode kit and faceplate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2120XPCALCalibration Kit
212050XPCa Hardness Consumables Kit
212050XPFReagentless CA Hardness Kit
2120XPEKElectrode Kit
2120XPEPElectronics Faceplate
Catalog number 2120XPCAL
Price (INR)
-
Type:
Calibration Kit
Use the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits with the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer. Accessories and kits include a calcium consumables kit, a reagent-less calcium hardness kit, a calcium hardness calibration kit with carrying case, a calcium electrode kit or a electronics faceplate.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
DescriptionCalcium Hardness Calibration Kit with Carrying Case - includes pipette, box of pipette tips, wash bottle, 2 oz. calcium standard 1 (191ppm), 2 oz. calcium standard 2 (1920ppm) and inlet filter
For Use With (Equipment)Orion™ 2120XP Calcium Hardness Analyzer
Includes0.5mL pipette, box of pipette tips, wash bottle, 2 oz. calcium standard 1, 2 oz calcium standard 2 and inlet filter
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeCalibration Kit
Unit SizeEach