Maximize coolant retention with these benchtop liquid nitrogen containers, which contain inner vessels made of borosilicate.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Includes
2123
2 L
Carrying Handle, Vented Secure Lid
2122
1 L
Carrying Handle, Vented Secure Lid
2124
4.5 L
Carrying Handle, Vented Secure Lid
2130TS1
also known as 2130
1.9 L
4 Options
Catalog number 2123
Price (INR)
-
Capacity (Metric):
2 L
Includes:
Carrying Handle, Vented Secure Lid
Maximize coolant retention with light-weight Thermo Scientific™ Thermo-Flask™ Benchtop Liquid Nitrogen Containers, which contain inner vessels of borosilicate and are perfect for everyday cryogenic storage use.