Sodium bromide, ACS reagent
Sodium bromide, ACS reagent
Sodium bromide, ACS reagent
Acros Organics

Sodium bromide, ACS reagent

CAS: 7647-15-6 | BrNa | 102.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 212671000
also known as 21267
HSN 28275110
Price (INR)
4,261.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,261.00
Each
Chemical Identifiers
CAS7647-15-6
IUPAC Namesodium bromide
Molecular FormulaBrNa
InChI KeyJHJLBTNAGRQEKS-UHFFFAOYSA-M
SMILES[Na+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or solid
Chloride (Cl)=<0.2 %
Barium (Ba)=<0.002 %
Calcium (Ca)=<0.002 %
Titration Argentometric>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only