Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Acros Organics

Ammonium titanyl oxalate monohydrate, 98%, pure

CAS: 10580-03-7 | C4H8N2O9Ti | 275.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
10 g
Packaging:
View all
Plastic bottle
Catalog number 212731000
HSN 29319090
Price (INR)
46,467.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
46,467.00
Each
Chemical Identifiers
CAS10580-03-7
IUPAC Namediammonium oxotitaniumbis(ylium) dioxalate
Molecular FormulaC4H8N2O9Ti
InChI KeyVBUNWMBOVJSSMH-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals
Titration with KMnO497.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only