DPBS, powder, calcium, magnesium
Actual product may vary
DPBS, powder, calcium, magnesium
DPBS, powder, calcium, magnesium
Gibco™

DPBS, powder, calcium, magnesium

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washingRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2130007450 L
21300017
also known as 21300-017
1 L
2130002510 x 1 L
213000581 x 10 L
Catalog number 21300074
Price (INR)
9,471.00
Each
Quantity:
50 L
Customize this product
Price (INR)
9,471.00
Each

Dulbecco's Phosphate-Buffered Saline (DPBS) is a balanced salt solution used for a variety of cell culture applications, such as washing cells before dissociation, transporting cells or tissue, diluting cells for counting, and preparing reagents. Formulations with calcium and magnesium are generally used as transport media or for reagent preparation. We offer a variety of Gibco™ DPBS formulations for a range of cell culture applications. Find the right formulation using the media selector tool.

This DPBS is modified as follows:

  • With
    • Calcium
    • Magnesium
  • Without
    • Phenol Red

The complete formulation is available.

For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the intended use may be a violation of local law.
Specifications
Chemical Name or MaterialDPBS
IncludesNo Sodium Pyruvate
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Osmolality260 - 310 mOsm/kg
Recommended StorageStorage conditions: 15-30° C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
For Use With (Application)Mammalian Cell Culture
Physical FormPowder
Product LineGibco
Quantity50 L
Solution TypeDulbecco's Phosphate Buffered Saline
pH7.3 - 7.5
Unit SizeEach

Frequently asked questions (FAQs)

Can I get a specific value for the conductivity for your DPBS products?

Sorry, we do not test the conductivity of our DPBS products and hence cannot provide a value or range.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Are your DPBS products DNase/RNase-free? Can I use them for RNA work?

Our DPBS products are for cell culture use and have not been tested for DNase/RNase activity. We do not recommend using these products for RNA work.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.