Selenourea, 98+%
Selenourea, 98+%
Selenourea, 98+%
Acros Organics

Selenourea, 98+%

CAS: 630-10-4 | CH4N2Se | 123.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 213170010
also known as 21317
HSN 29319090
Price (INR)
16,611.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
16,611.00
Each
Chemical Identifiers
CAS630-10-4
IUPAC Name[(C-selanylcarbonimidoyl)amino]-λ¹-selanyl
Molecular FormulaCH3N2Se
InChI KeyZUCFIOXGLBMWHV-UHFFFAOYSA-N
SMILESNC([Se])=N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals or needles
Assay>=98.0 %
Appearance (Color)White to pink to grey
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only