Streptavidin Protein, AP
Invitrogen™

Streptavidin Protein, AP

Protein
Have Questions?
Catalog NumberQuantity
213233 mg
Catalog number 21323
Price (INR)
53,328.00
Online offer
Ends: 31-Dec-2026
88,880.00
Save 35,552.00 (40%)
3 mg
Quantity:
3 mg
Price (INR)
53,328.00
Online offer
Ends: 31-Dec-2026
88,880.00
Save 35,552.00 (40%)
3 mg
Streptavidin for Western Blot, ELISA
Streptavidin is a tetrameric protein purified from Streptomyces avidinii that binds very tightly to the vitamin biotin with a Kd of ~ 10-14 mol/l. The high affinity recognition of biotin and biotinylated molecules has made streptavidin one of the most important components in diagnostics and laboratory kits.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Common NameStreptavidin
Concentration2 to 4 mg/mL
FormulationTBS with 50% glycerol, 0.1mM MgCl2, 0.1mM ZnCl2 and no preservative; pH 8
NameStreptavidin
Product TypeProtein
Purification MethodPurified
Quantity3 mg
Storage Requirements-20°C
ConjugateAlkaline Phosphatase (AP)
For Use With (Application)ELISA, Western Blot
FormLiquid
Regulatory StatusRUO
pH Range8
Unit Size3 mg
Contents & Storage
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.

Frequently asked questions (FAQs)

What is the relationship between specific activity units and International Units of activity?

There is no direct correlation or calculation between specific activity unit and International Unit (IU) values. International Units (IU) express a quantification of activity for the base amount of a substance in relation to an analogous reference standard with an internationally accepted unit of biological potency (i.e., IU/ng) that has been assigned based on an International Collaborative Study conducted by the World Health Organization (WHO). WHO Reference Standards are made available by the National Institute for Biological Standards and Control (NIBSC). Intended to simplify the comparison of activity of a substance obtained from different sources, IU measurements can vary as comparison methods are rarely the same between sources. A true direct comparison requires standardized methods of analysis in order to guarantee comparability of the substance’s activity in relation to its mass across sources.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Why can't I see the protein pellet in the vial for my lyophilized recombinant protein? Is this normal?

Gibco recombinant proteins are frequently formulated without carrier proteins or additives (e.g., BSA, HSA, sucrose, etc.) and Gibco PeproTech recombinant proteins don't contain a carrier protein. As a result, during lyophilization, the protein product may be deposited on the vial as a thin, and sometimes invisible film instead of a pellet. The size of the pellet, if any, is not directly related to the quantity of the recombinant protein in the vial. Our quality control procedures assure that each vial contains the correct amount of product.

To ensure complete recovery of protein product, before opening a vial of lyophilized recombinant protein, we recommend centrifuging it in a microcentrifuge for 20-30 seconds to drive any protein that may be lodged in the cap or on the side or the bottom of the vial. After reconstitution, you can confirm the presence of product protein by running a small amount on SDS-PAGE. In general, a protein band with expected size should be visible with as little as 10 ng of protein loaded on an acrylamide gel.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does a particular recombinant protein contain the wild-type or mutated amino acid sequence?

Protein sequence information is available on the individual product page, except for proprietary engineered proteins.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

What kind of tubes should I use for storing aliquots of reconstituted recombinant proteins?

In general, we recommend using polypropylene tubes for storing aliquots of reconstituted recombinant proteins. Specific information for appropriate storage and handling of many recombinant proteins can be found on the product pages.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Does each lot of recombinant protein have the same specific activity?

No, each lot can vary slightly with regards to its specific activity. The information on the data sheet for each product is provided as a guide. For lot-specific information, please please check the Certificate of Analysis or contact Technical Support.