MagnaBind™ Goat Anti-Mouse IgG
Thermo Scientific™

MagnaBind™ Goat Anti-Mouse IgG

Enable benchtop magnetic affinity purification with these ligand-coated (streptavidin, Protein A and G, GAM IgG, GAR IgG) magnetic particles and magnet separators.
Have Questions?
Catalog NumberQuantity
2135450 mL
Catalog number 21354
Price (INR)
39,288.00
線上優惠
Ends: 31-Dec-2026
65,480.00
Save 26,192.00 (40%)
Each
Quantity:
50 mL
Request bulk or custom format
Price (INR)
39,288.00
線上優惠
Ends: 31-Dec-2026
65,480.00
Save 26,192.00 (40%)
Each

Thermo Scientific™ MagnaBind™ Beads provide a convenient method for magnetic separation of antibodies, antigens, lectins, enzymes, nucleic acids and cells using affinity binding. To remove the MagnaBind Beads from the suspension, an external magnetic field is used.

MagnaBind Goat Anti-Mouse IgG Beads can be used to conveniently separate cells of interest from a cell mixture. The monoclonal or polyclonal antibody to the cell surface antigen is pre-incubated with the appropriate magnetic bead that is then incubated with the cell suspension.

Features of MagnaBind Goat Anti-Mouse IgG Beads:

  • Composition: Silanized iron oxide
  • Magnetization: 25-35EMU/g
  • Type of Magnetization: Superparamagnetic (no magnetic memory)
  • Surface Area: >100m2/g
  • Bead Size: 1-4μm diameter
  • Settling Rate: 4% in 30 minutes
  • Effective Density: 2.5g/mL
  • Number of Beads: 1 x 108 beads/mg
  • pH Stability: Aqueous solution, above pH 4.0
  • Concentration: ∼1 mg/mL

Applications:

  • Cell sorting using either positive or negative selection
  • Protein purification or immunoassays using either direct or indirect methods
For Research Use Only. Not for use in diagnostic procedures.
Specifications
ConcentrationSlurry: ∼1 mg/mL
DescriptionMagnaBind Goat Anti-Mouse IgG Beads
Ligand TypeAntibody
Product LineMagnaBind
Quantity50 mL
Volume (Metric)50 mL
ColorBrown
Diameter (Metric)1 to 4 μm
For Use With (Application)Affinity Purification
Product TypeAntibody Coated Bead
Unit SizeEach
Contents & Storage
Upon receipt store products at 4°C. Do not freeze products. Products are shipped at ambient temperature.