ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
ART™ Barrier Reload Insert Pipette Tips
Thermo Scientific™
ART™ Barrier Reload Insert Pipette Tips
Obtain accurate and consistent performance with these pipette tips, which seal when exposed to potential contaminants.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)ModelTip StyleFit for Purpose
2139-RIPK
also known as 2139-RI
10 μLART 10Micro PointSterile and PCR Ready
2065-RIPK
also known as 2065-RI
100 μLART 100Micro PointSterile and PCR Ready
2139-05-RIPK
also known as 2139-05-RI
10 μLART 10Low Retention, Micro PointSterile and PCR Ready
2149P-RIPK
also known as 2149P-RI
20 μLART 20PMicro PointSterile and PCR Ready
Catalog number 2139-RIPK
also known as 2139-RI
Price (INR)
7,399.00
Pack of 960
Volume (Metric):
10 μL
Model:
ART 10
Tip Style:
Micro Point
Fit for Purpose:
Sterile and PCR Ready
Price (INR)
7,399.00
Pack of 960
Obtain accurate and consistent performance with Thermo Scientific™ ART™ Barrier Reload Insert Pipette Tips, which seal when exposed to potential contaminants. The ART inserts are environmentally friendly, providing individually packaged blister tubs that are made out of recycled plastic, designed to be used with the reusable ART hinged racks and creating the ART Reload System. The tips from the reload inserts clip securely into the hinged racks for efficient reloading.
Features
  • Sterile tips are DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
  • Available in MicroPoint,extended length, low retention and SoftFit-L™ designed for use with Rainin™ LTS™ LiteTouch™ Pipettes
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Length (English)1.23 in.
Length (Metric)3.13cm
ModelART 10
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatRefill
Product LineART
SterilitySterile
Tip StyleMicro Point
Volume (Metric)10 μL
Unit SizePack of 960