Search
| Catalog Number | Volume (Metric) | Tip Style | Fit for Purpose |
|---|---|---|---|
| 2139-RTPK also known as 2139-RT | 10 μL | Standard | Sterile and PCR Ready |
| 2139-05-RTPK | 10 μL | Low Retention | Sterile and PCR Ready |
| 2149P-RTPK also known as 2149P-RT | 20 μL | Standard | Sterile and PCR Ready |
| 2069-RTPK also known as 2069-RT | 200 μL | Standard | Sterile and PCR Ready |
| 2069-05-RTPK also known as 2069-05-RT | 200 μL | Low Retention | Sterile and PCR Ready |
Perform applications accurately and consistently — without compromising protection — with the Thermo Scientific™ ART™ Barrier Reload System, which provides an environmentally responsible solution without compromising sterility. With offset wells that allow the tips to stack, this advanced system reduces waste, and saves benchtop and storage space. The ART portfolio is now more complete, making it easier than ever to find a tip to fit virtually any pipette or application in your laboratory.