ART REACH Barrier Tips, Extended Length
Molecular Bio Products Inc.
ART REACH Barrier Tips, Extended Length
Precise liquid handling with contamination protection, featuring extended reach and secure racks for efficiency.
Have Questions?
Catalog NumberVolume (Metric)
214010 μL
Catalog number 2140
Price (INR)
-
Volume (Metric):
10 μL
Precise liquid handling with contamination protection, featuring extended reach and secure racks for efficiency.
  • ART Reach Barrier Tips ensure accurate pipetting
  • Barrier filter prevents cross-contamination
  • Extended length design improves access to deep vessels
  • Hinged rack allows easy, one-handed operation
  • Sterile format suitable for sensitive applications
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
DescriptionART 10 REACH, Filtered, Sterile, Lift-off Lid Rack
Length (English)1.71 in.
Length (Metric)4.33cm
ModelART 10 REACH
Product TypePipette Tips
Quantity960 Tips
AutoclavableNot Autoclavable
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
For Use With (Equipment)Universal Fit
FormatLift-off Lid Rack
MaterialPolypropylene
Product LineART
SterilitySterile
Tip StyleExtended Length
Volume (Metric)10 μL
Unit SizeCase of 4800