ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
Thermo Scientific™
ART™ Pipette Tips, TipCycle Recycle Kit, Hinged Rack, Sterile
The pipette tip and rack mail-back recycling program provides a simple and effective solution for recycling single use plastics. The TipCycle Recycle Kit arrives with your case of pipette tips to collect used pipette tips and empty racks. Attach the pre-paid shipping label and return for recycling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Tip StyleModel
2140RC-HR10 μLExtended Length, Micro PointART 10 REACH
2079RC-05-HR1000 μLLow Retention, Extended Length, Micro PointART 1000
2079RC-HR1000 μLExtended Length, Micro PointART 1000
2140RC-05-HR10 μLLow Retention, Extended Length, Micro PointART 10 REACH
2179RC-05-HR1000 μLExtended Length, Micro PointART 1000L
Catalog number 2140RC-HR
Price (INR)
-
Volume (Metric):
10 μL
Tip Style:
Extended Length, Micro Point
Model:
ART 10 REACH
Unlike standard filter tips, Thermo Scientific™ ART™ Barrier Extended Length Pipette Tips in Hinged Racks seal when exposed to potential contaminants. The pipette tips from the reload inserts fit securely into the environmentally friendly, reusable hinged racks. The locking clasp prevents contamination by keeping remaining tips sterile and offers an easy, one-handed operation. Available in low retention, micropoint, extended length, and other specialty styles.
Features:
  • Recycle nonhazardous pipette tips, racks, and associated packaging with Tipcycle
  • Complete TipCycle Pipette Tip Recycling Kit Includes: 1 case of pipette tips, box liner, liner tie, FedEx return shipping label, waiver form, and instructions for return
  • Stackable hinged racks save space in your lab
  • Sterile tips are certified free from RNase, DNase and pyrogen
Recommended for:
  • Liquid Handling
  • Molecular Biology
  • PCR
  • Biotechnology
Specifications
Clean ClaimsDNase, RNase, DNA, ATP, PCR Inhibitor and Endotoxin Free
Length (English)1.71 in.
Length (Metric)4.33 cm
ModelART 10 REACH
Product TypeTipCycle Recycle Kit
AutoclavableNot Autoclavable
ColorNatural
FilteredFiltered
Fit for PurposeSterile and PCR Ready
FormatHinged Rack
Product LineART
SterilitySterile
Tip StyleExtended Length, Micro Point
Volume (Metric)10 μL
Unit SizeCase of 4800